Methoxyacetic acid 97+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Methoxyacetic acid

AC18677-0010 AC18677-0250 AC18677-2500
TS18677-0010EA 583 USD
TS18677-0010 TS18677-0250 TS18677-2500
Methoxyacetic acid 97+%
Methoxyacetic acid
Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.174 g/cm³
Flash Pt: 121 °C
MDL Number: MFCD00004308
CAS Number: 625-45-6
EINECS: 210-894-6
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4150 to 1.4180 (20°C, 589 nm)
Titration with NaOH 97.0 to 103.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR