Palladium(II) acetate 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Palladium diacetate, Palladium (II) acetate

AC44139-0010 AC44139-0050
TS44139-0010EA 155 USD
TS44139-0010 TS44139-0050
Palladium(II) acetate 99.9% (trace metals basis)
Palladium(II) acetate
Formula: C₄H₆O₄Pd
MW: 224.51 g/mol
Melting Pt: 205 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012453
CAS Number: 3375-31-3

Order Now

Specification Test Results

Appearance Orange to brown Powder
Assay 46.9 to 48.5 % (Pd)
Total trace metal impurities ≤1000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR