Methylmagnesium bromide 1 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC37738-1000 AC37738-8000
TS37738-1000EA 66 USD
TS37738-1000 TS37738-8000
Methylmagnesium bromide 1 M in THF, AcroSeal™
Methylmagnesium bromide
Formula: CH₃BrMg
MW: 119.24 g/mol
Density: 1 g/cm³
Flash Pt: –21 °C
Storage Temperature: Ambient
MDL Number: MFCD00000041
CAS Number: 75-16-1
EINECS: 200-844-1
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear to turbid gray to dark purple to brown-black Solution (May develop some turbidity or precipitate)
Titration Grignard 0.95 to 1.20 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR