Sodium metaborate tetrahydrate 98.5%, extra pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC21163-0025
AC21163-0100
AC21163-5000
TS21163-0025EA
135
USD
TS21163-0025
TS21163-0100
TS21163-5000
Sodium metaborate tetrahydrate 98.5%, extra pure
Sodium metaborate tetrahydrate
Formula:
BNaO₂·4H₂O Melting Pt: 57 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149205 CAS Number: 10555-76-7 EINECS: 231-891-6 |
Specification Test Results
Appearance | White Crystalline powder or crystals |
Titration with HCl | ≥98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...