Sodium ethoxide 21% in ethanol, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

AC42930-1000 AC42930-8000
TS42930-1000EA 42 USD
TS42930-1000 TS42930-8000
Sodium ethoxide 21% in ethanol, AcroSeal™
Sodium ethoxide
Formula: C₂H₅NaO
Boiling Pt: 91 °C
Density: 0.868 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Yellow to brown Solution (May darken on storage)
Titration with HCl 20.0 to 22.0 %
NaOH + Na2CO3 (as NaOH) ≤0.5 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR