Sodium ethoxide 21% in ethanol, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium ethylate, Sodium ethanolate

AC21322-0010 AC21322-1000 AC21322-2500
TS21322-0010EA 151 USD
TS21322-0010 TS21322-1000 TS21322-2500
Sodium ethoxide 21% in ethanol, pure
Sodium ethoxide
Formula: C₂H₅NaO
MW: 68.05 g/mol
Boiling Pt: 91 °C
Density: 0.868 g/cm³
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00012417
CAS Number: 141-52-6
EINECS: 205-487-5
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Yellow to brown Solution (May darken on storage)
Titration with HCl 20.0 to 22.0 %
NaOH + Na2CO3 (as NaOH) ≤0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR