Copper(II) phthalocyanine ∼95% (dye basis)

Supplier: Thermo Scientific Chemicals

Synonyms: 29H,31H-Phthalocyaninato-N29,N30,N31,N32 copper, Pigment Blue 15

AC26915-0250 AC26915-2500
TS26915-0250EA 48 USD
TS26915-0250 TS26915-2500
Copper(II) phthalocyanine ∼95% (dye basis)
Copper(II) phthalocyanine
Formula: C₃₂H₁₆CuN₈
MW: 576.08 g/mol
Melting Pt: 480 °C
MDL Number: MFCD00010719
CAS Number: 147-14-8
EINECS: 205-685-1
Merck Index: 12,02584

Order Now

Specification Test Results

Appearance Dark blue Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
Titration with Na2S2O3 ≥94 % (ex Cu)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR