Trimethyl orthoacetate 98+%

Supplier: Thermo Scientific Chemicals
Danger

AC21818-1000 AC21818-5000
TS21818-1000EA 54 USD
TS21818-1000 TS21818-5000
Trimethyl orthoacetate 98+%
Trimethyl orthoacetate
Formula: C₅H₁₂O₃
MW: 120.15 g/mol
Boiling Pt: 109 °C (1013 hPa)
Melting Pt: –58 °C
Density: 0.94 g/cm³
Flash Pt: 16 °C
MDL Number: MFCD00008477
CAS Number: 1445-45-0
EINECS: 215-892-9
UN: 3272
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3870 to 1.3900 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR