Gadolinium trinitrate hexahydrate 99.9% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

AC31676-0250 AC31676-1000
TS31676-0250EA 83 USD
TS31676-0250 TS31676-1000
Gadolinium trinitrate hexahydrate 99.9% (trace metals basis)
Gadolinium trinitrate hexahydrate
Formula: GdN₃O₉·6H₂O
Melting Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00149728
CAS Number: 19598-90-4
EINECS: 233-437-2
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Total conc. of imp.as REO ≤0.1 %
Gadolinium (Gd) 34 to 36 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR