Diethoxy(dimethyl)silane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC17463-1000
AC17463-5000
TS17463-1000EA
41
USD
TS17463-1000
TS17463-5000
Diethoxy(dimethyl)silane 97%
Diethoxy(dimethyl)silane
Formula:
C₆H₁₆O₂Si Boiling Pt: 114 °C (1013 hPa) Melting Pt: –70 °C Density: 0.86 g/cm³ Flash Pt: 11 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009068 CAS Number: 78-62-6 EINECS: 201-127-6 UN: 2380 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to slightly colored Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3805 to 1.3825 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...