Antimony(III) potassium oxitartrate trihydrate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R,R)-(+)-Antimony potassium tartrate trihydrate, (+)-Antimony potassium L-tartrate trihydrate, Tartar emetic, Antimony potassium L(+)-tartrate trihydrate, L(+)-Tartaric acid antimony potassium salt trihydrate, Potassium Antimony (III) Oxide Tartrate Trihydrate, (R,R)-(+)-Tartaric acid antimony potassium salt trihydrate, L(+)-Antimony potassium tartrate trihydrate, Potassium antimony(III) oxide tartrate trihydrate, Antimony potassium (+) tartrate trihydrate, Potassium antimony tartrate trihydrate, Potassium antimonyl tartrate sesquihydrate, Antimony potassium tartrate trihydrate, Potassium antimonyl tartrate trihydrate

AC22380-0025 AC22380-1000 AC22380-5000
TS22380-0025EA 573 USD
TS22380-0025 TS22380-1000 TS22380-5000
Antimony(III) potassium oxitartrate trihydrate 99+%
Antimony(III) potassium oxitartrate trihydrate
Formula: C₈H₄K₂O₁₂Sb₂·3H₂O
MW: 667.88 g/mol
Melting Pt: 100
Storage Temperature: Ambient
MDL Number: MFCD00167056
CAS Number: 28300-74-5
EINECS: 229-436-1
UN: 1551
ADR: 6.1 III
Merck Index: 13,00706

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥99.0 %
Loss on drying ≤2.7 % (105°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR