Bis(trimethylsilyl)carbodiimide 98%

Supplier: Thermo Scientific Chemicals
Warning

AC37513-0010 AC37513-0050 AC37513-0250
TS37513-0010EA 32 USD
TS37513-0010 TS37513-0050 TS37513-0250
Bis(trimethylsilyl)carbodiimide 98%
Bis(trimethylsilyl)carbodiimide
Formula: C₇H₁₈N₂Si₂
MW: 186.4 g/mol
Boiling Pt: 164 °C (1013 hPa)
Melting Pt: –23 °C
Density: 0.821 g/cm³
Flash Pt: 42 °C
MDL Number: MFCD00051538
CAS Number: 1000-70-0
EINECS: 213-673-2
UN: 1993

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4330 to 1.4350 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR