Trimethylsulphoxonium chloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Trimethyloxosulphonium chloride

AC38152-0010 AC38152-0050
TS38152-0010EA 39 USD
TS38152-0010 TS38152-0050
Trimethylsulphoxonium chloride 97%
Trimethylsulphoxonium chloride
Formula: C₃H₉ClOS
MW: 128.62
Melting Pt: 226…229 °C
Storage Temperature: Ambient
MDL Number: MFCD00011898
CAS Number: 5034-06-0

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR