Tris(triphenylphosphine)rhodium(I) chloride 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium

AC19457-0010 AC19457-0050 AC19457-2500
TS19457-0010EA 291 USD
TS19457-0010 TS19457-0050 TS19457-2500
Tris(triphenylphosphine)rhodium(I) chloride 99.99% (trace metals basis)
Tris(triphenylphosphine)rhodium(I) chloride
Formula: C₅₄H₄₅ClP₃Rh
MW: 925.23 g/mol
Melting Pt: 245…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00010016
CAS Number: 14694-95-2
EINECS: 238-744-5
Merck Index: 12,02226

Order Now

Specification Test Results

Appearance Brown Crystalline powder
Infrared spectrum Conforms
Total trace metal impurities ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR