Tris(triphenylphosphine)rhodium(I) chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Wilkinson's catalyst, Chlorotris(triphenylphosphine)rhodium(I), Tris(triphenylphosphine)chlororhodium

AC19042-0010 AC19042-0050
TS19042-0010EA 291 USD
TS19042-0010 TS19042-0050
Tris(triphenylphosphine)rhodium(I) chloride 98%
Tris(triphenylphosphine)rhodium(I) chloride
Formula: C₅₄H₄₅ClP₃Rh
MW: 925.23 g/mol
Melting Pt: 157
MDL Number: MFCD00010016
CAS Number: 14694-95-2
EINECS: 238-744-5
Merck Index: 12,02226

Order Now

Specification Test Results

Appearance Red-brown to brown Crystalline powder
Infrared spectrum Conforms
Gravimetry 10.8 to 11.3 % (Rh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR