Tetramethylammonium perchlorate 98%

Supplier: Thermo Scientific Chemicals
Danger

AC42056-0250 AC42056-1000
TS42056-0250EA 118 USD
TS42056-0250 TS42056-1000
Tetramethylammonium perchlorate 98%
Tetramethylammonium perchlorate
Formula: C₄H₁₂ClNO₄
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00043207
CAS Number: 2537-36-2
EINECS: 219-805-5
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Assay ≥97.5 % (on dry base)
Water 1 to 7 %
GC ≥99.0 %
Specific optical rotation +39° to +41° (20°C, 589 nm) (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR