Tetrakis(triphenylphosphine)platinum(0) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Tetrakis(triphenylphosphine)platinum

AC22396-0010 AC22396-0050
TS22396-0010EA 134 USD
TS22396-0010 TS22396-0050
Tetrakis(triphenylphosphine)platinum(0) 98%
Tetrakis(triphenylphosphine)platinum(0)
Formula: C₇₂H₆₀P₄Pt
MW: 1244.24 g/mol
Melting Pt: 159…160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010014
CAS Number: 14221-02-4
EINECS: 238-087-4

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Carbon (C) 67.8 to 71.2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR