Tetrakis(triphenylphosphine)nickel(0) 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC22398-0010
AC22398-0050
TS22398-0010EA
123
USD
TS22398-0010
TS22398-0050
Tetrakis(triphenylphosphine)nickel(0) 95%
Tetrakis(triphenylphosphine)nickel(0)
Formula:
C₇₂H₆₀NiP₄ Melting Pt: 122…124 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00010011 CAS Number: 15133-82-1 UN: 1325 ADR: 4.1,II |
Specification Test Results
Appearance | Rust brown to brown Powder or crystals |
Elemental Analysis | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...