Octadecyltrichlorosilane 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trichloro(stearyl)silane, Trichlorooctadecylsilane, Octadecyltrichlorosilane, ODTS, Trichloro(octadecyl)silane

AC14740-0250 AC14740-1000 AC14740-5000
TS14740-0250EA 51 USD
TS14740-0250 TS14740-1000 TS14740-5000
Octadecyltrichlorosilane 95%
Octadecyltrichlorosilane
Formula: C₁₈H₃₇Cl₃Si
MW: 387.94 g/mol
Boiling Pt: 361
Melting Pt: 22
Density: 0.98 g/cm³
Flash Pt: 189 °C
Storage Temperature: Ambient
MDL Number: MFCD00000484
CAS Number: 112-04-9
EINECS: 203-930-7
UN: 1800
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4585 to 1.4605 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR