Sodium perborate monohydrate 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC38641-0010
AC38641-0250
AC38641-2500
TS38641-0010EA
261
USD
TS38641-0010
TS38641-0250
TS38641-2500
Sodium perborate monohydrate 95%
Sodium perborate monohydrate
Formula:
BH₂NaO₄·1H₂O MW: 117.83 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00149230 CAS Number: 10332-33-9 EINECS: 239-172-9 UN: 3377 ADR: 5.1 III |
Specification Test Results
Appearance | White Crystalline powder |
Titration Iodometric | ≥94 % (active oxygen) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...