Sodium nitroprusside dihydrate 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium pentacyanonitrosylferrate(III) dihydrate, Sodium nitrosopentacyanoferrate (III) dihydrate, Sodium nitroferricyanide(III) dihydrate, Disodium pentacyanonitrosylferrate (III) dihydrate, Sodium nitroferricyanide dihydrate

AC21164-0250 AC21164-1000 AC21164-5000
TS21164-0250EA 56 USD
TS21164-0250 TS21164-1000 TS21164-5000
Sodium nitroprusside dihydrate 99+%
Sodium nitroprusside dihydrate
Formula: Na₂[Fe(CN)₅NO]·2H₂O
MW: 297.95 g/mol
Density: 1.72 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149192
CAS Number: 13755-38-9
EINECS: 238-373-9
UN: 1588
ADR: 6.1,III
Merck Index: 13,08722

Order Now

Specification Test Results

Appearance Red to dark red-brown Powder or crystals and/or chunks
Titration Argentometric ≥99.0 %
Chloride (Cl) ≤100 ppm
Sulfate (SO4) ≤100 ppm
Hexacyanoferrate III ≤0.01 %
Hexacyanoferrate II ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR