Methoxytrimethylsilane 96+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethyl(methoxy)silane

AC39247-0250 AC39247-1000 AC39247-2500
TS39247-0250EA 57 USD
TS39247-0250 TS39247-1000 TS39247-2500
Methoxytrimethylsilane 96+%
Methoxytrimethylsilane
Formula: C₄H₁₂OSi
MW: 104.22 g/mol
Boiling Pt: 49…51 °C (1013 hPa)
Density: 0.756 g/cm³
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00008344
CAS Number: 1825-61-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3660 to 1.3680 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR