Calcium formate 98%, pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formic acid calcium salt

AC40383-0010 AC40383-0025 AC40383-2500
TS40383-0010EA 233 USD
TS40383-0010 TS40383-0025 TS40383-2500
Calcium formate 98%, pure
Calcium formate
Formula: C₂H₂CaO₄
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00036108
CAS Number: 544-17-2
EINECS: 208-863-7
Merck Index: 12,01711

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %
Stabilizer 2 % CaCO3
Specific optical rotation +200° to +220° (20°C, 589 nm) (c=3, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR