Dipotassium tetraborate tetrahydrate 99.5%, extra pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Potassium tetraborate tetrahydrate

AC25957-0010 AC25957-0051 AC25957-2500
TS25957-0010EA 82 USD
TS25957-0010 TS25957-0051 TS25957-2500
Dipotassium tetraborate tetrahydrate 99.5%, extra pure
Dipotassium tetraborate tetrahydrate
Formula: B₄K₂O₇·4H₂O
MW: 305.49 g/mol
Melting Pt: 850 °C
Storage Temperature: Ambient
MDL Number: MFCD00149924
CAS Number: 12045-78-2
EINECS: 215-575-5

Order Now

Specification Test Results

Appearance White Powder
Titration with HClO4 ≥99.4 %
Chloride (Cl) ≤20 ppm
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤20 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR