N,O-Bis(trimethylsilyl)acetamide 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate

AC15646-0011 AC15646-0250 AC15646-1000
TS15646-0011EA 613 USD
TS15646-0011 TS15646-0250 TS15646-1000
N,O-Bis(trimethylsilyl)acetamide 95%
N,O-Bis(trimethylsilyl)acetamide
Formula: C₈H₂₁NOSi₂
MW: 203.43 g/mol
Boiling Pt: 71…73 °C (35 mmHg)
Melting Pt: –8 °C
Density: 0.82 g/cm³
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008270
CAS Number: 10416-59-8
EINECS: 233-892-7
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4160 to 1.4190 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR