Diammonium tetrachloropalladate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Ammonium tetrachloropalladate(II, Ammonium tetrachloropalladate(II), Ammonium palladium (II) chloride, Ammonium tetrachloropalladate (II)

AC19517-0020 AC19517-0100
TS19517-0020EA 244 USD
TS19517-0020 TS19517-0100
Diammonium tetrachloropalladate 98%
Diammonium tetrachloropalladate
Formula: H₈Cl₄N₂Pd
MW: 284.31 g/mol
Density: 2.17 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010884
CAS Number: 13820-40-1
EINECS: 237-498-6

Order Now

Specification Test Results

Appearance Green to red to brown Powder or crystals
Palladium (Pd) ≥36.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR