p-Phenylenediamine dihydrochloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

AC41748-0250 AC41748-1000
TS41748-0250EA 45 USD
TS41748-0250 TS41748-1000
p-Phenylenediamine dihydrochloride 99+%
p-Phenylenediamine dihydrochloride
Formula: C₆H₁₀Cl₂N₂
Melting Pt: 275 °C
Storage Temperature: Ambient
MDL Number: MFCD00013002
CAS Number: 624-18-0
EINECS: 210-834-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to yellow to purple-gray Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR