m-Phenylenediamine dihydrochloride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Benzene-1,3-diamine dihydrochloride, 1,3-Diaminobenzene dihydrochloride, 1,3-Phenylenediamine dihydrochloride, 1,3-Phenylenediammonium dichloride
AC21665-0250
AC21665-1000
TS21665-0250EA
55
USD
TS21665-0250
TS21665-1000
m-Phenylenediamine dihydrochloride 99%
m-Phenylenediamine dihydrochloride
Formula:
C₆H₁₀Cl₂N₂ MW: 181.06 g/mol Melting Pt: 277…278 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012975 CAS Number: 541-69-5 EINECS: 208-790-0 UN: 1673 ADR: 6.1 III |
Specification Test Results
Appearance | Light beige to light gray Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...