o-Phenylenediamine dihydrochloride 99+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzene-1,2-diamine dihydrochloride, 1,2-Phenylenediammonium dichloride, 1,2-Diaminobenzene dihydrochloride, 1,2-Phenylenediamine dihydrochloride

AC21848-0250 AC21848-1000
TS21848-0250EA 187 USD
TS21848-0250 TS21848-1000
o-Phenylenediamine dihydrochloride 99+%
o-Phenylenediamine dihydrochloride
Formula: C₆H₁₀Cl₂N₂
MW: 181.06 g/mol
Melting Pt: 205 (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012966
CAS Number: 615-28-1
EINECS: 210-418-7
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Light pink to purple Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR