(Trifluoromethyl)trimethylsilane 0.5 M in THF

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: trimethyl(trifluoromethyl)silane

AC36832-0050 AC36832-0250 AC36832-1000
TS36832-0050EA 33 USD
TS36832-0050 TS36832-0250 TS36832-1000
(Trifluoromethyl)trimethylsilane 0.5 M in THF
(Trifluoromethyl)trimethylsilane
Formula: C₄H₉F₃Si
MW: 142.2 g/mol
Boiling Pt: 40 °C (760 mmHg)
Density: 0.895 g/cm³
Flash Pt: –20 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145454
CAS Number: 81290-20-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.45 to 0.55 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR