(Trifluoromethyl)trimethylsilane 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: trimethyl(trifluoromethyl)silane

AC27972-0010 AC27972-0050 AC27972-0250
TS27972-0010EA 62 USD
TS27972-0010 TS27972-0050 TS27972-0250
(Trifluoromethyl)trimethylsilane 99%
(Trifluoromethyl)trimethylsilane
Formula: C₄H₉F₃Si
Boiling Pt: 54…55 °C (760 mmHg)
Density: 0.95 g/cm³
Flash Pt: –17 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00145454
CAS Number: 81290-20-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3290 to 1.3320 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR