Vinyltrimethylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethyl(vinyl)silane

AC20033-0250 AC20033-1000
TS20033-0250EA 128 USD
TS20033-0250 TS20033-1000
Vinyltrimethylsilane 97%
Vinyltrimethylsilane
Formula: C₅H₁₂Si
MW: 100.24 g/mol
Boiling Pt: 55 °C (1013 hPa)
Density: 0.68 g/cm³
Flash Pt: –24 °C
MDL Number: MFCD00008606
CAS Number: 754-05-2
EINECS: 212-042-9
UN: 1993

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3895 to 1.3915 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR