N,N-Dimethyl-1,1,1-trimethylsilylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (Dimethylamino)trimethylsilane, Trimethylsilyldimethylamine, N,N-Dimethyltrimethylsilylamine, TMSDMA, N-(Trimethylsilyl)dimethylamine, N,N-Dimethylaminotrimethylsilane, Pentamethylsilylamine

AC20997-0100 AC20997-0500
TS20997-0100EA 47 USD
TS20997-0100 TS20997-0500
N,N-Dimethyl-1,1,1-trimethylsilylamine 97%
N,N-Dimethyl-1,1,1-trimethylsilylamine
Formula: C₅H₁₅NSi
MW: 117.27 g/mol
Boiling Pt: 84 °C (1013 hPa)
Density: 0.732 g/cm³
Flash Pt: –19 °C
Storage Temperature: Ambient
MDL Number: MFCD00008297
CAS Number: 2083-91-2
EINECS: 218-222-3
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3960 to 1.3980 (20°C, 589 nm)
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR