N,N-Diethyl-1,1,1-trimethylsilylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Trimethylsilyldiethylamine, (Diethylamino)trimethylsilane, N,N-Diethylaminotrimethylsilane, N,N-Diethyltrimethylsilylamine, N-(Trimethylsilyl)diethylamine, TMSDEA

AC15569-0050 AC15569-0250 AC15569-1000
TS15569-0050EA 23 USD
TS15569-0050 TS15569-0250 TS15569-1000
N,N-Diethyl-1,1,1-trimethylsilylamine 98+%
N,N-Diethyl-1,1,1-trimethylsilylamine
Formula: C₇H₁₉NSi
MW: 145.32 g/mol
Boiling Pt: 125…126 °C (760 mmHg)
Melting Pt: –10 °C
Density: 0.76 g/cm³
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00009040
CAS Number: 996-50-9
EINECS: 213-637-6
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4103 to 1.4123 (20°C, 589 nm)
GC ≥98.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR