(Chloromethyl)trimethylsilane 98+%

Supplier: Thermo Scientific Chemicals
Danger

AC19476-0250 AC19476-1000
TS19476-0250EA 79 USD
TS19476-0250 TS19476-1000
(Chloromethyl)trimethylsilane 98+%
(Chloromethyl)trimethylsilane
Formula: ClH₂CSi(CH₃)₃
MW: 122.67 g/mol
Boiling Pt: 98 °C (1013 hPa)
Density: 0.879 g/cm³
Flash Pt: –2 °C
MDL Number: MFCD00000878
CAS Number: 2344-80-1
EINECS: 219-058-5
UN: 1993

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4165 to 1.4185 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR