Iron(III) acetylacetonate 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(2,4-pentanedionato-O,O')iron (III), Ferric acetylacetonate, Iron (III) 2,4-pentanedionate, Iron (III) tris(2,4-pentanedionato-O,O')

AC11913-0010 AC11913-0250 AC11913-1000 AC11913-2500
TS11913-0010EA 393 USD
TS11913-0010 TS11913-0250 TS11913-1000 TS11913-2500
Iron(III) acetylacetonate 99+%
Iron(III) acetylacetonate
Formula: C₁₅H₂₁FeO₆
MW: 353.18 g/mol
Boiling Pt: 100
Melting Pt: 180…183 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00000020
CAS Number: 14024-18-1
EINECS: 237-853-5

Order Now

Specification Test Results

Appearance Dark orange to dark red Powder or crystals
Infrared spectrum Conforms
Titration Iodometric ≥99.0 %
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR