Platinum(II) acetylacetonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC36935-0010
AC36935-0050
AC36935-0250
TS36935-0010EA
200
USD
TS36935-0010
TS36935-0050
TS36935-0250
Platinum(II) acetylacetonate 98%
Platinum(II) acetylacetonate
Formula:
C₁₀H₁₄O₄Pt Melting Pt: 250…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000028 CAS Number: 15170-57-7 EINECS: 239-223-5 |
Specification Test Results
Appearance | Yellow to yellow-green Powder |
Infrared spectrum | Conforms |
Assay | ≥97.5 % (ex Pt) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...