Cobalt(II) acetylacetonate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC21064-0500
AC21064-2500
TS21064-0500EA
93
USD
TS21064-0500
TS21064-2500
Cobalt(II) acetylacetonate 99%
Cobalt(II) acetylacetonate
Formula:
C₁₀H₁₄CoO₄ Melting Pt: 165…170 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000014 CAS Number: 14024-48-7 EINECS: 237-855-6 |
Specification Test Results
Appearance | Purple Crystalline powder |
Infrared spectrum | Conforms |
Assay | 98.5 to 101.5 % (ex Co) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...