Manganese(III) acetylacetonate 97%

Supplier: Thermo Scientific Chemicals
Warning

AC34398-0050 AC34398-0250 AC34398-1000
TS34398-0050EA 20 USD
TS34398-0050 TS34398-0250 TS34398-1000
Manganese(III) acetylacetonate 97%
Manganese(III) acetylacetonate
Formula: C₁₅H₂₁MnO₆
Melting Pt: 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00000023
CAS Number: 14284-89-0
EINECS: 238-188-3

Order Now

Specification Test Results

Appearance Black Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥96 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR