Chromium(III) acetylacetonate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(2,4-pentanedionato-O,O')chromium (III), Chromium (III) 2,4-pentanedionate, Chromium (III) tris(2,4-pentanedionato-O,O')

AC19213-0050 AC19213-1000 AC19213-5000
TS19213-0050EA 37 USD
TS19213-0050 TS19213-1000 TS19213-5000
Chromium(III) acetylacetonate 97%
Chromium(III) acetylacetonate
Formula: C₁₅H₂₁CrO₆
MW: 349.32 g/mol
Boiling Pt: 340 °C (760 mmHg)
Melting Pt: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD00000015
CAS Number: 21679-31-2
EINECS: 244-526-0

Order Now

Specification Test Results

Appearance Brown to purple Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Assay 14.4 to 15.3 % (Cr)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR