Molybdenum pentachloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
AC19708-0100
AC19708-1000
TS19708-0100EA
48
USD
TS19708-0100
TS19708-1000
Molybdenum pentachloride 95%
Molybdenum pentachloride
Formula:
Cl₅Mo MW: 273.2 g/mol Boiling Pt: 268 Melting Pt: 194 |
MDL Number:
MFCD00003467 CAS Number: 10241-05-1 EINECS: 233-575-3 UN: 2508 ADR: 8 III |
Specification Test Results
Appearance | Blue to green-black to black Crystalline powder or crystals |
Titration Argentometric | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...