Cerium(III) sulphate 99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dicerium trisulphate

AC37866-0500 AC37866-2500
TS37866-0500EA 77 USD
TS37866-0500 TS37866-2500
Cerium(III) sulphate 99.9% (REO, rare earth oxide basis)
Cerium(III) sulphate
Formula: Ce₂O₁₂S₃
MW: 568.42
Density: 2.89
CAS Number: 13454-94-9
EINECS: 236-644-6

Order Now

Specification Test Results

Appearance White to off-white Powder or crystals
Titration Complexometric 46.0 to 50.0 % (Ce)
Total conc. of imp.as REO ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR