Tetraethylammonium nitrate 99%

Supplier: Thermo Scientific Chemicals
Danger

AC42031-0100 AC42031-0250
TS42031-0100EA 130 USD
TS42031-0100 TS42031-0250
Tetraethylammonium nitrate 99%
Tetraethylammonium nitrate
Formula: C₈H₂₀N₂O₃
MW: 192.26
Melting Pt: >240 °C
Density: 1.1622
Storage Temperature: Ambient
MDL Number: MFCD00041978
CAS Number: 1941-26-0
EINECS: 217-725-5
UN: 1479
ADR: 5.1 II

Order Now

Specification Test Results

Appearance White to yellowish-white Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR