Dodecyltrimethylammonium bromide 99%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide
AC40931-0250
AC40931-1000
AC40931-5000
TS40931-0250EA
109
USD
TS40931-0250
TS40931-1000
TS40931-5000
Dodecyltrimethylammonium bromide 99%, pure
Dodecyltrimethylammonium bromide
Formula:
C₁₅H₃₄BrN MW: 308.35 g/mol Melting Pt: 246 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00011767 CAS Number: 1119-94-4 EINECS: 214-290-3 UN: 3077 ADR: 9,III |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥98.5 % |
Solubility | (10 % in water) Clear to slightly hazy |
Color of solution | ≤100 APHA (10% aq. soln.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...