Bismuth chloride oxide 97%, pure
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bismuth oxychloride, Bismuth (III) oxychloride, Bismuth(III) chloride oxide
AC45384-0010
AC45384-0500
AC45384-2500
TS45384-0010EA
508
USD
TS45384-0010
TS45384-0500
TS45384-2500
Bismuth chloride oxide 97%, pure
Bismuth chloride oxide
Formula:
BiClO MW: 260.43 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010895 CAS Number: 7787-59-9 |
Specification Test Results
Appearance | White to off-white Powder |
Assay | 77.0 to 82.5 % (Bi) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...