Digallium trioxide 99.99+% (trace metals basis)

Supplier: Thermo Scientific Chemicals

AC20110-0050 AC20110-0500
TS20110-0050EA 67 USD
TS20110-0050 TS20110-0500
Digallium trioxide 99.99+% (trace metals basis)
Digallium trioxide
Formula: Ga₂O₃
MW: 187.44 g/mol
Melting Pt: 1740 °C
Density: 5.88 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011020
CAS Number: 12024-21-4
EINECS: 234-691-7

Order Now

Specification Test Results

Appearance White Crystalline powder
Total trace metal impurities ≤0.010 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR