Potassium hydrogen difluoride 99+%, extra pure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Potassium hydrogen fluoride, Potassium bifluoride

AC21923-0025 AC21923-0250 AC21923-5000
TS21923-0025EA 273 USD
TS21923-0025 TS21923-0250 TS21923-5000
Potassium hydrogen difluoride 99+%, extra pure
Potassium hydrogen difluoride
Formula: HF₂K
MW: 78.1 g/mol
Melting Pt: 239
MDL Number: MFCD00011403
CAS Number: 7789-29-9
EINECS: 232-156-2
UN: 1811
ADR: 8 II
Merck Index: 12,07771

Order Now

Specification Test Results

Appearance White to light gray Crystalline powder or crystals
Titration with NaOH ≥99.0 %
Sulfate (SO4) ≤500 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR