Benzeneselenenyl bromide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylselenyl bromide, Phenyl hypobromoselenoite, Bromoselenobenzene, Phenylselenenyl bromide

AC22306-0050 AC22306-0250
TS22306-0050EA 72 USD
TS22306-0050 TS22306-0250
Benzeneselenenyl bromide 98%
Benzeneselenenyl bromide
Formula: (C₆H₅)SeBr
MW: 235.97 g/mol
Boiling Pt: 229
Melting Pt: 60…62 °C
MDL Number: MFCD00000047
CAS Number: 34837-55-3
EINECS: 252-238-1
UN: 2928
ADR: 6.1 II

Order Now

Specification Test Results

Appearance Dark brown to black Crystals
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR