Cobalt(II) carbonate hydrate

Supplier: Thermo Scientific Chemicals
Danger

AC19210-0010 AC19210-0500 AC19210-2500
TS19210-0010EA 507 USD
TS19210-0010 TS19210-0500 TS19210-2500
Cobalt(II) carbonate hydrate
Cobalt(II) carbonate hydrate
Formula: CCoO₃·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00149654
CAS Number: 57454-67-8
EINECS: 208-169-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Pink to red to violet Powder
Titration Complexometric ≥45.5 % (Cobalt)
Cadmium (Cd) ≤50 ppm
Copper (Cu) ≤100 ppm
Iron (Fe) ≤100 ppm
Insoluble matter ≤0.05 % (in HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR