Cobalt(II) bromide hydrate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cobalt dibromide hydrate, Cobaltous bromide hydrate

AC31854-1000 AC31854-5000
TS31854-1000EA 135 USD
TS31854-1000 TS31854-5000
Cobalt(II) bromide hydrate
Cobalt(II) bromide hydrate
Formula: Br₂Co·H₂O
MW: 236.76 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150208
CAS Number: 85017-77-2
EINECS: 232-166-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Violet Crystals
Titration Argentometric 17.0 to 20.3 % (Co)
Water 24.7 to 36.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR